Delmhorst® JX-30 Moisture Meters
Features the Delmhorst EDGE™ App with expanded wood species, customizable settings, and the ability to share data from any location
  • Compatible with Delmhorst EDGE™ app via Bluetooth® connectivity
  • Measurement range of 6 to 60% wood (Douglas Fir)
  • 74 species included plus 123 additional available through the EDGE™ App
  • Large, custom display with auto backlight for easy reading under all conditions
  • Wood temperature correction (°F/°C)
  • Pins/electrode correction (insulated or non-insulated)
  • Alarm set point alerts to user-selected MC level
  • On-screen statistics – High, Low, Average, Standard Deviation, View all readings
  • Internal calibration check
  • Integral contact pins max 5/16” penetration (use 2498/A-100)
The JX-30 meters are ideal tools for woodworkers, furniture and cabinet manufacturers, sawmills, wood flooring contractors, and any other wood product related field. They are packaged in a robust and ergonomically designed ABS case to provide a premium, tactile feel, and intuitive user interface with dashboard-like display. This meter enhances your ability to ensure precise readings and export data from anywhere, including geotags and timestamps with Bluetooth® connectivity.
116,000.00 - ₹152,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Electrode Type Moisture Range Wood Scale (Max) (%) Description
Availability Pricing
Delmhorst JX-30 Digital Pin Moisture Meter with Bluetooth® Connectivity 5980071 Mfr # JX-30W/CS
COMPARE
None included 60 Digital Pin Moisture Meter with Bluetooth® Connectivity
₹116,000.00INR / Each
Delmhorst JX-30 Digital Pin Moisture Meter with Bluetooth® Connectivity, Wood Package 5980072 Mfr # JX-30/P01
COMPARE
26-ES hammer electrode 60 Digital Pin Moisture Meter with Bluetooth® Connectivity, Wood Package
₹152,000.00INR / Each