Delmhorst® JX-20 Moisture Meters
Bringing lumber/woodworking, flooring professionals and inspectors professional-grade, accurate, and versatile tools to address moisture issues and perform jobs with complete confidence
  • Measurement range of 6 to 60% wood (Douglas Fir)
  • 74 species included
  • Large, custom display with auto backlight for easy reading under all conditions
  • Wood temperature correction (°F/°C)
  • Pins/electrode correction (insulated or non-insulated)
  • Alarm set point alerts to user-selected MC level
  • On-screen statistics – High, Low, Average, Standard Deviation, View all readings
  • Internal calibration check
  • Integral contact pins max 5/16” penetration (use 2498/A-100)
The JX-20 meters are ideal tools for woodworkers, furniture and cabinet manufacturers, sawmills, wood flooring contractors and any other wood product related field. They are packaged in a robust and ergonomically designed ABS case to provide a premium, tactile feel, and intuitive user interface with dashboard-like display.
92,900.00 - ₹130,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Electrode Type Moisture Range Wood Scale (Max) (%) Description
Availability Pricing
Delmhorst JX-20 Digital Pin Moisture Meter 5980069 Mfr # JX-20W/CS
COMPARE
None included 60 Digital Pin Moisture Meter
₹92,900.00INR / Each
Delmhorst JX-20 Digital Pin Moisture Meter, Wood Package 5980070 Mfr # JX-20/P01
COMPARE
26-ES hammer electrode 60 Digital Pin Moisture Meter, Wood Package
₹130,000.00INR / Each