Delmhorst® FX-30 Agriculture Moisture Meters
Optimize moisture control for hay, hemp, hops, and more with the FX-30, now featuring Bluetooth® connectivity for easy data sharing and customization via the Delmhorst EDGE® app
  • Wide moisture range settings for six commodities, with temperature correction
  • Bluetooth® connectivity for seamless sync and data transfer to EDGE® app
  • Customizable, color-coded LED alerts and alarm set points through EDGE®
  • Saves up to 100 readings with High, Low, Average, and Standard Deviation display
  • Sturdy ABS case with large, tactile buttons and ergonomic E-Z grip
The FX-30 Moisture Meter is the go-to tool for quality control and moisture monitoring in a variety of commodities, including hay, hops, hemp, and tobacco. With a robust, ergonomic ABS case and a clear dashboard-like display, the FX-30 offers precision at every step. This advanced tool comes with Bluetooth® connectivity, allowing users to pair with the Delmhorst® EDGE® app to set alarms, view color-coded LED indicators, and save up to 100 readings for a comprehensive view of moisture levels. Compatible with all Delmhorst® electrodes and prods, the FX-30 ensures durability, accuracy, and seamless sharing options wherever you work.
84,400.00 - ₹134,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type Measurement Type Description
Availability Pricing
Delmhorst FX-30 Digital Agriculture Meter with Bluetooth® 5980079 Mfr # FX-30
COMPARE
None included Hay (6-40%), Hops (8-23%), Hemp (7-34%), Tobacco (8-37%), Brazil Nuts (6-25%), Dates (6-25%) Digital Agriculture Meter with Bluetooth®
₹84,400.00INR / Each
Delmhorst FX-30 Digital Agriculture Meter with Bluetooth®, Value Package 5980080 Mfr # FX-30/1235
COMPARE
1235 10" prod Hay (6-40%), Hops (8-23%), Hemp (7-34%), Tobacco (8-37%), Brazil Nuts (6-25%), Dates (6-25%) Digital Agriculture Meter with Bluetooth®, Value Package
₹100,000.00INR / Each
Delmhorst FX-30 Digital Agriculture Meter with Bluetooth®, Value 18" Package 5980081 Mfr # FX-30/1235/18
COMPARE
1235/18 18" prod Hay (6-40%), Hops (8-23%), Hemp (7-34%), Tobacco (8-37%), Brazil Nuts (6-25%), Dates (6-25%) Digital Agriculture Meter with Bluetooth®, Value 18" Package
₹104,000.00INR / Each
Delmhorst FX-30 Digital Agriculture Meter with Bluetooth®, Deluxe Package 5980082 Mfr # FX-30/P01
COMPARE
830-2 10" prod and 831 windrow probe Hay (6-40%), Hops (8-23%), Hemp (7-34%), Tobacco (8-37%), Brazil Nuts (6-25%), Dates (6-25%) Digital Agriculture Meter with Bluetooth®, Deluxe Package
₹133,000.00INR / Each
Delmhorst FX-30 Digital Agriculture Meter with Bluetooth®, Deluxe 18" Package 5980083 Mfr # FX-30/P02
COMPARE
830-3 18" prod and 831 windrow probe Hay (6-40%), Hops (8-23%), Hemp (7-34%), Tobacco (8-37%), Brazil Nuts (6-25%), Dates (6-25%) Digital Agriculture Meter with Bluetooth®, Deluxe 18" Package
₹134,000.00INR / Each
Delmhorst FX-30 Digital Agriculture Meter with Bluetooth®, Windrow Package 5980084 Mfr # FX-30/P03
COMPARE
831 windrow probe Hay (6-40%), Hops (8-23%), Hemp (7-34%), Tobacco (8-37%), Brazil Nuts (6-25%), Dates (6-25%) Digital Agriculture Meter with Bluetooth®, Windrow Package
₹119,000.00INR / Each