Delmhorst® FX-20 Hay Moisture Meter
Stay ahead of moisture issues and maintain the quality of your hay with the FX-20 Moisture Meter, designed for accurate and reliable moisture monitoring in all conditions
  • Wide moisture range (6-40% for hay) and internal calibration for trustworthy readings
  • Large, custom display with auto-backlight for easy reading in any light
  • Modern ABS case with E-Z grip and tactile buttons for comfort and durability
  • Saves and displays statistics for up to 100 readings, including High, Low, Average, and Standard Deviation
  • Customizable alarm set point to alert at user-selected moisture levels
The FX-20 Moisture Meter is a must-have for hay producers and quality control experts, delivering precise moisture readings to preserve hay quality from field to storage. Packaged in a rugged, ergonomically designed ABS case, the FX-20 combines durability with an intuitive interface, including an easy-to-read dashboard-like display, on-screen statistics, and customizable alarms. Compatible with all Delmhorst® prods and electrodes, this meter is the ultimate tool for ensuring optimal moisture levels, preventing mold, and protecting feed value.
68,400.00 - ₹118,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type Measurement Type Description
Availability Pricing
Delmhorst FX-20 Digital Hay Moisture Meter 5980073 Mfr # FX-20
COMPARE
None included Hay 6-40% Digital Hay Moisture Meter
₹68,400.00INR / Each
Delmhorst FX-20 Digital Hay Moisture Meter, Value Package 5980074 Mfr # FX-20/1235
COMPARE
1235 10" prod Hay 6-40% Digital Hay Moisture Meter, Value Package
₹83,400.00INR / Each
Delmhorst FX-20 Digital Hay Moisture Meter, Value 18" Package 5980075 Mfr # FX-20/1235/18
COMPARE
1235/18 18" prod Hay 6-40% Digital Hay Moisture Meter, Value 18" Package
₹86,500.00INR / Each
Delmhorst FX-20 Digital Hay Moisture Meter, Deluxe Package 5980076 Mfr # FX-20/P01
COMPARE
830-2 10" prod and 831 windrow probe Hay 6-40% Digital Hay Moisture Meter, Deluxe Package
₹117,000.00INR / Each
Delmhorst FX-20 Digital Hay Moisture Meter, Deluxe 18" Package 5980077 Mfr # FX-20/P02
COMPARE
830-3 18" prod and 831 windrow probe Hay 6-40% Digital Hay Moisture Meter, Deluxe 18" Package
₹118,000.00INR / Each
Delmhorst FX-20 Digital Hay Moisture Meter, Windrow Package 5980078 Mfr # FX-20/P03
COMPARE
831 windrow probe Hay 6-40% Digital Hay Moisture Meter, Windrow Package
₹102,000.00INR / Each