Delmhorst® BDX-30 Moisture Meters
The powerful BDX-30 brings all the design elements and functionality of the BDX-20 and introduces Bluetooth® integration with the Delmhorst EDGE™ app.
  • Compatible with Delmhorst EDGE™ app via Bluetooth® connectivity
  • Download up to 12 species/materials into the meter from the 37 available in the app
  • 3 measurement scales: 6 to 60% wood (Douglas Fir), 0.1 to 6% drywall, and 0 to 100 reference scale
  • Large, custom display with auto backlight makes for easy reading under all conditions
  • Wood temperature correction (°F/°C)
  • Alarm set point alerts to a user-selected MC level
  • On-screen statistics – High, Low, Average, Standard Deviation, View all readings
  • Internal calibration check
  • Pins/electrode correction (insulated or non-insulated)
The BDX-30 brings today’s technology from the field to the office, making the most challenging project easier, more efficient, and more accurate. This meter enhances your ability to ensure precise readings and export data from anywhere, including geotags and timestamps. With the EDGE™ app, users can also select from 37 species and materials.
119,000.00 - ₹193,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Electrode Type Moisture Range Wood Scale (Max) (%) Description
Availability Pricing
Delmhorst BDX-30 Digital Pin Moisture Meter with Bluetooth® Connectivity 5980065 Mfr # BDX-30W/CS
COMPARE
None included 60 Digital Pin Moisture Meter with Bluetooth® Connectivity
₹119,000.00INR / Each
Delmhorst BDX-30 Digital Pin Moisture Meter with Bluetooth® Connectivity, Behind the Wall Package 5980066 Mfr # BDX-30/P01
COMPARE
21-E electrode 60 Digital Pin Moisture Meter with Bluetooth® Connectivity, Behind the Wall Package
₹141,000.00INR / Each
Delmhorst BDX-30 Digital Pin Moisture Meter with Bluetooth® Connectivity, Wood Flooring Package 5980067 Mfr # BDX-30/P02
COMPARE
26-ES hammer electrode 60 Digital Pin Moisture Meter with Bluetooth® Connectivity, Wood Flooring Package
₹152,000.00INR / Each
Delmhorst BDX-30 Digital Pin Moisture Meter with Bluetooth® Connectivity, Contractor/EIFS Package 5980068 Mfr # BDX-30/P03
COMPARE
26-ES and 21-E electrodes 60 Digital Pin Moisture Meter with Bluetooth® Connectivity, Contractor/EIFS Package
₹193,000.00INR / Each