Delmhorst® BDX-20 Moisture Meters
Durable, precise, and easy to use with a custom display and specialized measurement scales for reliable readings on every job
  • 3 measurement scales: 6 to 60% wood (Douglas Fir), 0.1 to 6% drywall, and 0 to 100 reference scale
  • Large, custom display with auto backlight makes for easy reading under all conditions
  • Wood temperature correction (°F/°C)
  • Alarm set point alerts to a user-selected MC level
  • On-screen statistics – High, Low, Average, Standard Deviation, View all readings
  • Internal calibration check
  • Pins/electrode correction (insulated or non-insulated)
The BDX-20 is ideal for use in the building trades, water damage restoration, and inspection applications. This rugged meter has an ergonomic design with an easy grip handle and large tactile buttons for comfort and usability. Equipped with a custom LCD display and intuitive user interface, users can quickly obtain accurate readings.
96,200.00 - ₹139,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Electrode Type Moisture Range Wood Scale (Max) (%) Description
Availability Pricing
Delmhorst BDX-20 Digital Pin Moisture Meter 5980063 Mfr # BDX-20W/CS
COMPARE
None included 60 Digital Pin Moisture Meter
₹96,200.00INR / Each
Delmhorst BDX-20 Digital Pin Moisture Meter, Behind the Wall Package 5980064 Mfr # BDX-20/P01
COMPARE
21-E (3" penetration) 60 Digital Pin Moisture Meter, Behind the Wall Package
₹119,000.00INR / Each
Delmhorst BDX-20 Digital Pin Moisture Meter, 26-ES Slide Hammer Package 5980093 Mfr # BDX-20/P02
COMPARE
26-ES slide hammer 60 Digital Pin Moisture Meter, 26-ES Slide Hammer Package
₹132,000.00INR / Each
Delmhorst BDX-20 Digital Pin Moisture Meter, EIFS Package 5980094 Mfr # BDX-20/P03
COMPARE
21-E, HP-1 hole punch 60 Digital Pin Moisture Meter, EIFS Package
₹139,000.00INR / Each