Delmhorst Advanced RDM3 Moisture Meters
The wide assortment of meters and electrodes gives the flexibility to choose the right combination for your application
  • Specific meters are available for wood or paper applications
  • Measures moisture through built-in expand capabilities with optional pin electrodes
These handheld moisture meters are packed with features and are ideal for anyone who needs to collect and analyze key statistical information related to moisture. Pin-type moisture meters are the standard around the world as they provide the best way to identify surface and core moisture content, minimizing defects and improving quality control.
Store up to 1400 readings and view statistics for each job number; data can be downloaded via optional software. Display the stored readings or show the average, standard deviation, minimum, and maximum values. Date/time stamp on all stored readings helps with quality control programs. Adjustable audible alarm alerts you when a preselected moisture value is reached.
RDM3 Meter has been designed for monitoring moisture in wood. The contact pins mounted on top of the meter provide 5⁄16" penetration for testing wood up to 1" (2.5 cm) thick. The meter measures wood over the range of 6 to 60%, and include built-in corrections for 71 individual wood species.
RDM3/P Meter is ideal for collecting and analyzing key statistical information related to paper. The contact pins mounted on top of the meter provide 5⁄16" (0.8 cm) penetration for testing paper tubes or corrugated stock. Several optional external probes are available for measuring percent of moisture content on a moving web.
182,000.00 - ₹250,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type Description
Availability Pricing
Delmhorst RDM3/PKG Advanced Rdm-3P Moisture Meter, Expanded Package 5982043 Mfr # RDM3/PKG
COMPARE
1 9V (included) Pin Advanced Rdm-3P Moisture Meter, Expanded Package
₹205,000.00INR / Each
Delmhorst RDM3/PLUS Advanced Rdm-3P Moisture Meter, Expanded/Data Logging Package 5982044 Mfr # RDM3/PLUS
COMPARE
1 9V (included) Pin Advanced Rdm-3P Moisture Meter, Expanded/Data Logging Package
₹250,000.00INR / Each
Delmhorst RDM-3PW/CS Advanced Rdm-3P Moisture Meter, Basic Package 5982056 Mfr # RDM-3PW/CS
COMPARE
1 9V (included) Pin Advanced Rdm-3P Moisture Meter, Basic Package
₹182,000.00INR / Each
Delmhorst RDM-3P/PKG Advanced Rdm-3P Moisture Meter, Data Logging Package 5982057 Mfr # RDM-3P/PKG
COMPARE
1 9V (included) Pin Advanced Rdm-3P Moisture Meter, Data Logging Package
₹228,000.00INR / Each