Crucibles, Quartz
Quartz crucible satisfies high-temperature requirements up to 1922°F (1050°C)—perfect for muffle furnace ashing procedures. Quartz material ensures chemical purity for subsequent analysis. Crucible is useful for determining trace metals in environmental samples.
67,800.00 - ₹103,000.00INR / Case of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottom Outside Diameter (mm) Capacity (mL) Cap Material
Availability Pricing
DWK Life Sciences (Kimble) Quartz Crucible and Cover, 50 mL, 49 mm OD X 50 mm H 3452650 Mfr # 319010-0050
COMPARE
49 50 Quartz
₹103,000.00INR / Case of 1
DWK Life Sciences (Kimble) Quartz Crucible, no cover, 50 mL, 49 mm OD X 50 mm H 3452652 Mfr # 319011-0050
COMPARE
49 50
₹67,800.00INR / Case of 1