CR Magnetics DC and AC-DC Current and Voltage Transducers
  • APPLICATIONS
  • Battery chargers and systems
  • DC motor drives
  • Power supply management
  • Mobile applications
  • Power sensing
  • Inverter and multi-frequency drives (AC/DC)
  • Multi-mode ground paths with AC and DC (AC/DC)
  • Panel or DIN rail mounts
The Series 52 DC Current Transducers are designed to output a DC signal which is proportional to a DC current input signal. These devices are for direct current only, targeted towards general DC current sensing.
The Series 54 AC/DC Current Transducers provide a bipolar output that proportionally reflects the input bipolar current. These devices are specifically designed to be used in applications where multi-mode current sensing is required.
The Series 53 DC Voltage Transducers provide an output DC signal that is proportional to the input DC Voltage. These devices are focused to provide an easy solution to DC Voltage instrumentation needs.
1 PRODUCT OPTIONS
Compareitem specifications
Basic Accuracy (%) Max Input (VDC) Max Output (VDC)
Availability Pricing
CR Magnetics CR4110-25 True RMS AC Current Transducer, 0~5 VDC Output 6848074 Mfr # CR4110-25
COMPARE
0.5%
₹69,600.00INR / Each