CPC® MC Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved, Chrome-Plated Brass
Provides quick and secure connection and disconnection of fluid lines
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly, even with gloved hands
  • Withstands higher pressure and temperature
  • Mix and match termination sizes for added flexibility
  • Compatible with PMC acetal and PMC12 polypropylene couplings
The CPC (Colder) MC series quick-disconnect hose barb body and insert fittings provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. An ergonomic thumb latch allows you to safely connect and disconnect the fittings with the use of one hand, even with gloves on.
These chrome-plated brass fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures.
1,300.00 - ₹4,800.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) MC Quick-Disconnect Fitting, Hose Barb Body, Chrome-Plated Brass, Non-Valved, 1/8" ID; 1/Each 5013573 Mfr # MC1702
COMPARE
1/8 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹4,400.00INR / Each
CPC (Colder) MC Quick-Disconnect Fitting, Hose Barb Body, Chrome-Plated Brass, Non-Valved, 1/4" ID; 1/Each 5013574 Mfr # MC1704
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹4,800.00INR / Each
CPC (Colder) MC Quick-Disconnect Fitting, Hose Barb Insert, Chrome-Plated Brass, Non-Valved, 1/8" ID; 1/Each 5013576 Mfr # MC2202
COMPARE
1/8 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹1,300.00INR / Each
CPC (Colder) MC Quick-Disconnect Fitting, Hose Barb Insert, Chrome-Plated Brass, Non-Valved, 1/4" ID; 1/Each 5013577 Mfr # MC2204
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹1,400.00INR / Each