CPC® (Colder) SMC Quick-Disconnect Fittings, Threaded Body
Provides more secure alternative to luer-type fittings
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Free coupling rotation eliminates kinked tubing
  • Ideal for tubing sizes from 1/16" to 1/8" ID
The CPC (Colder) SMC Series quick-disconnect hose barb body and insert fittings offer a twist-to-connect design, providing a reliable and more secure alternative to luer-type fittings. This design allows tubing to rotate freely when connected, preventing both kinked tubing and accidental disconnection during use. The fittings can withstand pressures from vacuum to 100 psi (6.9 bar).
330.00 - ₹3,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Threaded Body, Acetal, Valved, 1/8" NPT(M); 1/Ea 4852735 Mfr # SMPTD02
COMPARE
Acetal Quick-Disconnect Threaded Body, Valved 1/8
₹3,300.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Threaded Body, Acetal, Non-Valved, 1/8" NPT(M); 1/Ea 4852736 Mfr # SMPT02
COMPARE
Acetal Quick-Disconnect Threaded Body, Non-Valved 1/8
₹450.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Threaded Body, PP, Non-Valved, 1/8" NPT(M); 1/Ea 5013681 Mfr # SMPT0212
COMPARE
Polypropylene Quick-Disconnect Threaded Body, Non-Valved 1/8
₹330.00INR / Each