CPC® (Colder) SMC Quick-Disconnect Fittings, Hose Barb Body & Insert
Provides more secure alternative to luer-type fittings
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Free coupling rotation eliminates kinked tubing
  • Ideal for tubing sizes from 1/16” to 1/8” ID
The CPC (Colder) SMC Series quick-disconnect hose barb body and insert fittings offer a twist-to-connect design, providing a reliable and more secure alternative to luer-type fittings. This design allows tubing to rotate freely when connected, preventing both kinked tubing and accidental disconnection during use. The fittings can withstand pressures from vacuum to 100 psi (6.9 bar),
The fittings are available in acetal and polypropylene. The acetal fittings operate effectively within a temperature range of -40°F to 180°F (-40°C to 82°C). The polypropylene fittings operate effectively within a temperature range of 32 to 160 °F (0 to 71 °C).
370.00 - ₹15,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Non-Valved, 1/8" ID; 50/Pk 1541404 Mfr # SMM02/50 PK
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹15,000.00INR / Pkg of 50
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 1/8" ID; 1/Ea 4852730 Mfr # SMF02
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Body, Non-Valved
₹560.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Body, PP, Non-Valved, 1/16" ID; 1/Ea 4852731 Mfr # SMF0112
COMPARE
1/16 Polypropylene Quick-Disconnect Hose Barb Body, Non-Valved
₹790.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Insert, PP, Non-Valved, 1/16" ID; 1/Ea 4852732 Mfr # SMM0112
COMPARE
1/16 Polypropylene Quick-Disconnect Hose Barb Insert, Non-Valved
₹1,000.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 1/16" ID; 1/Ea 4852733 Mfr # SMFD01
COMPARE
1/16 Acetal Quick-Disconnect Hose Barb Body, Valved
₹3,300.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 1/8" ID; 1/Ea 4852734 Mfr # SMFD02
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Body, Valved
₹3,300.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 1/16" ID; 1/Ea 5013676 Mfr # SMF01
COMPARE
1/16 Acetal Quick-Disconnect Hose Barb Body, Non-Valved
₹370.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Body, PP, Non-Valved, 1/8" ID; 1/Ea 5013677 Mfr # SMF0212
COMPARE
1/8 Polypropylene Quick-Disconnect Hose Barb Body, Non-Valved
₹390.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID; 1/Ea 5013678 Mfr # SMM01
COMPARE
1/16 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹430.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Non-Valved, 1/8" ID; 1/Ea 5013679 Mfr # SMM02
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹370.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Hose Barb Insert, PP, Non-Valved, 1/8" ID; 1/Ea 5013680 Mfr # SMM0212
COMPARE
1/8 Polypropylene Quick-Disconnect Hose Barb Insert, Non-Valved
₹560.00INR / Each
Showing 1- 11 of 11