CPC® (Colder) PMCD Quick-Disconnect Fittings, Threaded Body & Insert, Valved
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
The CPC (Colder) PMCD Series quick-disconnect threaded body and insert fittings have a 1/8" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
The ergonomic thumb latch design allows for effortless, one-handed connection and disconnection, even when wearing gloves. Integral terminations reduce leak points, shorten assemblies, and facilitate faster installations. These fittings can withstand pressures ranging from vacuum to 120 psi (8.3 bar).
1,500.00 - ₹45,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Threaded Body, Polypropylene, Valved, 1/4" NPT(M); 25/PK 4901493 Mfr # PMCD100412/25 PK
COMPARE
Polypropylene Quick-Disconnect Threaded Body, Valved 1/4
₹45,000.00INR / Pkg of 25
CPC (Colder) PMCD Quick-Disconnect Fitting, Threaded Body, Acetal, Valved, 1/8" NPT(M) 5013643 Mfr # PMCD1002
COMPARE
Acetal Quick-Disconnect Threaded Body, Valved 1/8
₹1,500.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Threaded Body, Polypropylene, Valved, 1/8" NPT(M) 5013644 Mfr # PMCD100212
COMPARE
Polypropylene Quick-Disconnect Threaded Body, Valved 1/8
₹1,900.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Threaded Body, Acetal, Valved, 1/4" NPT(M) 5013645 Mfr # PMCD1004
COMPARE
Acetal Quick-Disconnect Threaded Body, Valved 1/4
₹1,500.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Threaded Body, Polypropylene, Valved, 1/4" NPT(M) 5013646 Mfr # PMCD100412
COMPARE
Polypropylene Quick-Disconnect Threaded Body, Valved 1/4
₹2,000.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, 1/8" NPT(M) 5013668 Mfr # PMCD2402
COMPARE
Acetal Quick-Disconnect Threaded Insert, Valved 1/8
₹1,600.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, 1/4" NPT(M) 5013669 Mfr # PMCD2404
COMPARE
Acetal Quick-Disconnect Threaded Insert, Valved 1/4
₹1,600.00INR / Each
Showing 1- 7 of 7