CPC® (Colder) PMCD Quick-Disconnect Fittings, PTF Body & Insert, Valved
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
The CPC (Colder) PLCD Series quick disconnect ferruleless polytube (PTF) body and insert fittings, designed for semi-rigid tubing, have a 1/8" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings can withstand pressures from vacuum to 120 psi (8.3 bar).
1,400.00 - ₹51,900.00INR / Pkg of 25
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Flow Pattern Thread Size PTF OD (port 1) (InchShort)
Availability Pricing
CPC (Colder) PMCD12 Quick-Disconnect Fittings, PTF Insert, Valved, Polypropylene, 1/4" OD; 25PK 4901471 Mfr # PMCD200412/25 PK
COMPARE
Polypropylene Straight 0.25
₹46,300.00INR / Pkg of 25
CPC (Colder) PMCD Quick-Disconnect Fittings, PTF Body, Panel Mount, Valved, Acetal, 1/4" OD; 25/PK 4901563 Mfr # PMCD1204 25/PK
COMPARE
Acetal Straight 0.25
₹51,900.00INR / Pkg of 25
CPC (Colder) PMCD Quick-Disconnect Fittings, PTF Body, Panel Mount, Valved, Acetal, 1/4" OD 5013647 Mfr # PMCD1204
COMPARE
Acetal Straight 0.25
₹2,300.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fittings, PTF Body, Valved, Acetal, 1/4" OD 5013648 Mfr # PMCD1304
COMPARE
Acetal Straight 0.25
₹1,600.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fittings, PTF Insert, Valved, Acetal, 1/4" OD 5013659 Mfr # PMCD2004
COMPARE
Acetal Straight 0.25
₹1,400.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fittings, PTF Insert, Valved, Polypropylene, 1/4" OD 5013660 Mfr # PMCD200412
COMPARE
Polypropylene Straight 0.25
₹2,100.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fittings, PTF Insert, Elbow, Valved, Polypropylene, 1/4" OD 5013661 Mfr # PMCD2104
COMPARE
Acetal Elbow 0.25
₹2,200.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fittings, PTF Insert, Panel Mount, Valved, Polypropylene, 1/4" OD 5013670 Mfr # PMCD4004
COMPARE
Acetal Straight 0.25
₹2,800.00INR / Each
Showing 1- 8 of 8