CPC® (Colder) PMCD Quick-Disconnect Fittings, Hose Barb Body & Inset, Valved
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
The CPC (Colder) PMCD Series quick-disconnect hose barb body and insert fittings have a 1/8" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
The ergonomic thumb latch design allows for effortless, one-handed connection and disconnection, even when wearing gloves. Integral terminations reduce leak points, shorten assemblies, and facilitate faster installations. These fittings can withstand pressures ranging from vacuum to 120 psi (8.3 bar).
1,400.00 - ₹32,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Flow Pattern
Availability Pricing
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Valved, 1/8" ID; 25/PK 4901356 Mfr # PMCD2202/25 PK
COMPARE
1/8 Acetal Straight
₹32,000.00INR / Pkg of 25
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Polypropylene, Valved, 1/16" ID 5013649 Mfr # PMCD160112
COMPARE
1/16 Polypropylene Straight
₹2,900.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Polypropylene, Valved, 1/8" ID 5013650 Mfr # PMCD160212
COMPARE
1/8 Polypropylene Straight
₹2,600.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Acetal, Valved, 3/16" ID 5013651 Mfr # PMCD1603
COMPARE
3/16 Acetal Straight
₹2,200.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Acetal, Valved, 1/4" ID 5013652 Mfr # PMCD1604
COMPARE
1/4 Acetal Straight
₹2,200.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 1/8" ID 5013653 Mfr # PMCD1702
COMPARE
1/8 Acetal Straight
₹1,600.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 1/8" ID 5013654 Mfr # PMCD170212
COMPARE
1/8 Polypropylene Straight
₹2,600.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 3/16" ID 5013655 Mfr # PMCD1703
COMPARE
3/16 Acetal Straight
₹1,600.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 1/4" ID 5013656 Mfr # PMCD1704
COMPARE
1/4 Acetal Straight
₹1,600.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 1/4" ID 5013657 Mfr # PMCD170412
COMPARE
1/4 Polypropylene Straight
₹2,600.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Valved, 1/8" ID 5013662 Mfr # PMCD2202
COMPARE
1/8 Acetal Straight
₹1,400.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 1/8" ID 5013663 Mfr # PMCD220212
COMPARE
1/8 Polypropylene Straight
₹1,800.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Valved, 3/16" ID 5013664 Mfr # PMCD2203
COMPARE
3/16 Acetal Straight
₹1,500.00INR / Each
CPC (Colder) PMCD Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Valved, 1/4" ID 5013665 Mfr # PMCD2204
COMPARE
1/4 Acetal Straight
₹1,400.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 1/4" ID 5013666 Mfr # PMCD220412
COMPARE
1/4 Polypropylene Straight
₹2,000.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Polypropylene, Valved, 1/8" ID 5013667 Mfr # PMCD230212
COMPARE
1/75 Polypropylene Elbow
₹2,700.00INR / Each
Showing 1- 16 of 16