CPC® (Colder) PMC Quick-Disconnect Fittings, Threaded Body & Insert, Non-Valved, Acetal
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/16” to 1/4” ID
The CPC (Colder) PMC Series quick-disconnect threaded body and insert fittings have a 1/8" flow size, making them suitable for a wide range of general-purpose and chemical applications. Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions.
The ergonomic thumb latch design allows for effortless, one-handed connection and disconnection, even when wearing gloves. Integral terminations reduce leak points, shorten assemblies, and facilitate faster installations. These fittings can withstand pressures ranging from vacuum to 120 psi (8.3 bar) and operate effectively within a temperature range of -40 to 180 °F (-40 to 82 °C).
530.00 - ₹1,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) PMC Quick-Disconnect Fitting, Threaded Body, Acetal, Non-Valved, 1/8" NPT(M); 1/Each 5013631 Mfr # PMC1002
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valved 1/8
₹1,100.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Threaded Body, Acetal, Non-Valved, 1/4" NPT(M); 1/Each 5013632 Mfr # PMC1004
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valved 1/4
₹1,200.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Threaded Insert, Acetal, Non-Valved, 1/8" NPT(M); 1/Each 5013641 Mfr # PMC2402
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valved 1/8
₹530.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Threaded Insert, Acetal, Non-Valved, 1/4" NPT(M); 1/Each 5013642 Mfr # PMC2404
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valved 1/4
₹580.00INR / Each