CPC® (Colder) PMC Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
The CPC (Colder) PMC Series quick-disconnect hose barb body and insert fittings have a 1/8" flow size, making them suitable for a wide range of general-purpose and chemical applications.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
The ergonomic thumb latch design allows for effortless, one-handed connection and disconnection, even when wearing gloves. Integral terminations reduce leak points, shorten assemblies, and facilitate faster installations. These fittings can withstand pressures ranging from vacuum to 120 psi (8.3 bar).
300.00 - ₹12,600.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Flow Pattern
Availability Pricing
CPC (Colder) PMC12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Non-Valved, 1/4" ID; 25/Pk 1846067 Mfr # PMC220412 25/PK
COMPARE
1/4 Polypropylene Straight
₹12,600.00INR / Pkg of 25
CPC (Colder) PMC Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Acetal, Non-Valved, 1/8" ID; 1/Each 5013633 Mfr # PMC1602
COMPARE
1/8 Acetal Straight
₹1,800.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 1/8" ID; 1/Each 5013634 Mfr # PMC1702
COMPARE
1/8 Acetal Straight
₹840.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Non-Valved, 1/8" ID; 1/Each 5013636 Mfr # PMC2202
COMPARE
1/8 Acetal Straight
₹300.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Non-Valved, 1/4" ID; 1/Each 5013637 Mfr # PMC2204
COMPARE
1/4 Acetal Straight
₹330.00INR / Each
CPC (Colder) PMC12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Non-Valved, 1/4" ID; 1/Each 5013638 Mfr # PMC220412
COMPARE
1/4 Polypropylene Straight
₹580.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Acetal, Non-Valved, 1/8" ID; 1/Each 5013639 Mfr # PMC2302
COMPARE
1/8 Acetal Elbow
₹650.00INR / Each
CPC (Colder) PMC Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Acetal, Non-Valved, 1/4" ID; 1/Each 5013640 Mfr # PMC2304
COMPARE
1/4 Acetal Elbow
₹640.00INR / Each
Showing 1- 8 of 8