CPC® (Colder) PLCD Quick-Disconnect Fittings, Threaded Body & Insert, Valved
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLCD Series quick disconnect threaded body and insert fittings have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings can withstand pressures from vacuum to 120 psi (8.3 bar).
1,500.00 - ₹37,500.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) 1/4" VALVED NPT THREAD COUPLING BODIES ACETAL WITH BUNA-N O-RING 25/PK 1565003 Mfr # PLCD10004
COMPARE
₹37,500.00INR / Pkg of 25
CPC (Colder) PLCD12 Quick-Disconnect Fittings,  Threaded Insert, Polypropylene, Valved, 1/4" NPT(M); 1/Each 5013608 Mfr # PLCD1000412
COMPARE
Polypropylene Quick-Disconnect Threaded Body, Valve 1/4
₹2,100.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fittings,  Threaded Body, Acetal, Valved, 3/8" NPT(M); 1/Each 5013609 Mfr # PLCD10006
COMPARE
Acetal Quick-Disconnect Threaded Body, Valve 3/8
₹1,500.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fittings,  Threaded Insert, Acetal, Valved, 1/4" NPT(M); 1/Each 5013627 Mfr # PLCD24004
COMPARE
Acetal Quick-Disconnect Threaded Insert, Valve 1/4
₹1,500.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fittings,  Threaded Insert, Acetal, Valved, 3/8" NPT(M); 1/Each 5013628 Mfr # PLCD24006
COMPARE
Acetal Quick-Disconnect Threaded Insert, Valve 3/8
₹1,500.00INR / Each