CPC® (Colder) PLCD Quick-Disconnect Fittings, PTF Body & Insert, Valved, Acetal & Polypropylene
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLCD Series quick disconnect ferruleless polytube (PTF) body and insert fittings, designed for semi-rigid tubing, have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings can withstand pressures from vacuum to 120 psi (8.3 bar).
1,700.00 - ₹37,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Thread Size PTF OD (port 1) (InchShort)
Availability Pricing
CPC (Colder) PLCD Quick-Disconnect Fitting, PTF Insert, Acetal, Valved, 3/8" OD; 25PK 4901459 Mfr # PLCD20006/25 PK
COMPARE
Acetal Quick-Disconnect PTF Insert, Valve 0.375
₹37,800.00INR / Pkg of 25
CPC (Colder) PLCD Quick-Disconnect Fitting, PTF Body, Acetal, Valved, 1/4" OD 5013610 Mfr # PLCD13004
COMPARE
Acetal Quick-Disconnect PTF Body, Valve 0.25
₹2,200.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fitting, PTF Insert, Acetal, Valved, 1/4" OD 5013617 Mfr # PLCD20004
COMPARE
Acetal Quick-Disconnect PTF Insert, Valve 0.25
₹1,700.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fitting, PTF Insert, Acetal, Valved, 3/8" OD 5013618 Mfr # PLCD20006
COMPARE
Acetal Quick-Disconnect PTF Insert, Valve 0.375
₹1,700.00INR / Each
CPC (Colder) PLCD12 Quick-Disconnect Fitting, PTF Insert, Polypropylene, Valved, 3/8" OD 5013619 Mfr # PLCD2000612
COMPARE
Polypropylene Quick-Disconnect PTF Insert, Valve 0.375
₹2,600.00INR / Each