CPC® (Colder) PLCD Quick-Disconnect Fittings, JG® Body & Insert, Valved, Acetal
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLCD Series quick disconnect JG body and insert fittings have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings can operate in temperatures from -40 to 180 °F (-40 to 82 °C) and withstand pressures from vacuum to 120 psi (8.3 bar).
2,100.00 - ₹46,400.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
CPC (Colder) PLCD Quick-Disconnect Fitting, JG Insert, Acetal, Valved, 1/4" OD; 25/PK 4901343 Mfr # PLCD29004/25 PK
COMPARE
Straight Quick-Disconnect JG Push-to-Connect Insert, Valve 1/4
₹46,400.00INR / Pkg of 25
CPC (Colder) PLCD Quick-Disconnect Fitting, JG Body, Acetal, Valved, 1/4" OD 5013611 Mfr # PLCD14004
COMPARE
Straight Quick-Disconnect JG Push-to-Connect Body, Valve 1/4
₹2,500.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fitting, JG Insert, Acetal, Valved, 1/4" OD 5013629 Mfr # PLCD29004
COMPARE
Straight Quick-Disconnect JG Push-to-Connect Insert, Valve 1/4
₹2,100.00INR / Each
CPC (Colder) PLCD Quick-Disconnect Fitting, JG Insert, Acetal, Valved, 3/8" OD 5013630 Mfr # PLCD29006
COMPARE
Straight Quick-Disconnect JG Push-to-Connect Insert, Valve 3/8
₹2,300.00INR / Each