CPC® (Colder) PLC Quick-Disconnect Fittings, Threaded Insert, Non-Valved, Acetal
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLC Series quick disconnect threaded insert fittings have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings operate efficiently in temperatures ranging from -40 to 180 °F (-40 to 82 °C) and withstand pressures from vacuum to 120 psi (8.3 bar).
370.00 - ₹12,900.00INR / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) PLC Quick-Disconnect Fitting, Threaded Insert, Acetal, Non-Valved, 1/4" NPT(M); 25/PK 4901285 Mfr # PLC24004/25 PK
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valved 1/4
₹12,900.00INR / Pkg of 25
CPC (Colder) PLC Quick-Disconnect Fitting, Threaded Insert, Acetal, Non-Valved, 1/4" NPT(M) 5013605 Mfr # PLC24004
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valved 1/4
₹520.00INR / Each
CPC (Colder) PLC Quick-Disconnect Fitting, Threaded Insert, Acetal, Non-Valved, 3/8" NPT(M) 5013606 Mfr # PLC24006
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valved, 3/8
₹370.00INR / Each