CPC® (Colder) PLC Quick-Disconnect Fittings, JG® Body & Insert, Non-Valved, Acetal
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLC Series quick disconnect JG body and insert fittings have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings operate efficiently in temperatures ranging from -40 to 180 °F (-40 to 82 °C) and withstand pressures from vacuum to 120 psi (8.3 bar).
1,400.00 - ₹2,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Push-to-Connect Tube OD (port 1) (in)
Availability Pricing
CPC (Colder) PLC Quick-Disconnect Fitting, JG Body, Acetal, Non-Valved, 1/4" OD; 1/Each 5013598 Mfr # PLC14004
COMPARE
Straight Quick-Disconnect JG Body, Non-Valved 1/4
₹2,400.00INR / Each
CPC (Colder) PLC Quick-Disconnect Fitting, JG® Insert, Acetal, Non-Valved, 1/4" OD; 1/Each 5013607 Mfr # PLC29004
COMPARE
Straight Quick-Disconnect JG Insert, Non-Valved 1/4
₹1,400.00INR / Each