CPC® (Colder) PLC Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved, Acetal & Polypropylene
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from 1/4” to 3/8” ID
The CPC (Colder) PLC Series quick disconnect hose barb body and insert fittings have a 1/4" flow size, making them suitable for a wide range of general-purpose and chemical applications. Constructed from durable acetal thermoplastic, these fittings offer high strength, rigidity across a broad temperature range, and resistance to many mild chemical solutions. Alternatively, polypropylene versions provide enhanced chemical resistance, making them ideal for harsher environments.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings can withstand pressures from vacuum to 120 psi (8.3 bar).
230.00 - ₹24,600.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) PLC Quick-Disconnect Fittings, Hose Barb Body, Acetal, Non-Valved, 1/4" ID; 25/PK 4901284 Mfr # PLC17004/25 PK
COMPARE
1/4 Acetal Quick-Disconnect Hose Barb Body, Non-Valved
₹24,600.00INR / Pkg of 25
CPC (Colder) PLC Quick-Disconnect Fittings, Hose Barb Body, Panel Mount, Acetal, Non-Valved, 1/4" ID; 1/Each 5013599 Mfr # PLC16004
COMPARE
1/4 Acetal Quick-Disconnect Hose Barb Body, Panel Mount, Non-Valved
₹2,800.00INR / Each
CPC (Colder) PLC Quick-Disconnect Fittings, Hose Barb Body, Acetal, Non-Valved, 1/4" ID; 1/Each 5013600 Mfr # PLC17004
COMPARE
1/4 Acetal Quick-Disconnect Hose Barb Body, Non-Valved
₹940.00INR / Each
CPC (Colder) PLC Quick-Disconnect Fittings, Hose Barb Body, Acetal, Non-Valved, 3/8" ID; 1/Each 5013601 Mfr # PLC17006
COMPARE
3/8 Acetal Quick-Disconnect Hose Barb Body, Non-Valved
₹980.00INR / Each
CPC (Colder) PLC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/4" ID; 1/Each 5013602 Mfr # PLC22004
COMPARE
1/4 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹230.00INR / Each
CPC (Colder) PLC12 Quick-Disconnect Fittings, Hose Barb Insert, Polypropylene, Non-Valved, 1/4" ID; 1/Each 5013603 Mfr # PLC2200412
COMPARE
1/4 Polypropylene Quick-Disconnect Hose Barb Insert, Non-Valved
₹520.00INR / Each
CPC (Colder) PLC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 3/8" ID; 1/Each 5013604 Mfr # PLC22006
COMPARE
3/8 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹260.00INR / Each
Showing 1- 7 of 7