CPC® (Colder) NS6 Quick-Disconnect Fittings, Hose Barb Body & Insert, Valved, Polypropylene
Designed for a pressure-activated release without leaks
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Chemically resistant
  • Ideal for tubing sizes from 3/8” to 1/2” ID
The CPC (Colder) NS6 Series quick-disconnect hose barb body and insert fittings are designed with non-spill valves to prevent spills, reduce downtime, and improve operator safety. These lightweight, compact fittings offer excellent chemical resistance and user-friendly operation.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fittings operate efficiently in temperatures ranging from 32 to 160 °F (0 to 71 °C) and withstand pressures from vacuum to 120 psi (8.3 bar).
3,300.00 - ₹4,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) NS6 Quick-Disconnect Fittings, Hose Barb Body, Polypropylene, Valved, 1/2" ID; 1/Each 5013595 Mfr # NS6D17008
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Body, Valve
₹4,800.00INR / Each
CPC (Colder) NS6 Quick-Disconnect Fittings, Hose Barb Insert, Polypropylene, Valved, 1/2" ID; 1/Each 5013596 Mfr # NS6D22008
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Insert, Valve
₹3,300.00INR / Each