CPC® (Colder) MPX® Quick-Disconnect Fittings, Plug
Connect to a female coupling body to stop the flow
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
CPC® (Colder) MPX® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Use a plug with a body to seal off the fluid pathway.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
28,400.00 - ₹82,200.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
CPC (Colder) MPX® Quick-Disconnect Fitting, Plug, Polycarbonate; 25/Pk 5011323 Mfr # MPX30003MB25
COMPARE
Purple tint Polycarbonate Quick-Disconnect Fitting, Plug, Polycarbonate; 25/Pk
₹28,400.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Plug, Polysulfone; 25/Pk 5011325 Mfr # MPX30039MB25
COMPARE
Amber tint Polysulfone Quick-Disconnect Fitting, Plug, Polysulfone; 25/Pk
₹82,200.00INR / Pkg of 25