CPC® (Colder) MPX® Quick-Disconnect Fittings, Hose Barb Insert, Non-Valved
Connect to a coupling body to quickly create a sterile fluid pathway
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a female coupling body for easy media transfer
  • Mix and match termination sizes for added flexibility
  • 0.5" nominal flow path size
CPC® (Colder) MPX® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Simply use a cap to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
24,700.00 - ₹74,200.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) MPX® Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Non-Valved, 3/8" ID; 25/Pk 5011314 Mfr # MPX22603MB25
COMPARE
3/8 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Non-Valved, 3/8" ID; 25/Pk
₹24,700.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 3/8" ID; 25/Pk 5011316 Mfr # MPX22639MB25
COMPARE
3/8 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 3/8" ID; 25/Pk
₹74,200.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Non-Valved, 1/2" ID; 25/Pk 5011318 Mfr # MPX22803MB25
COMPARE
1/2 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Non-Valved, 1/2" ID; 25/Pk
₹24,800.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 1/2" ID; 25/Pk 5011320 Mfr # MPX22839MB25
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 1/2" ID; 25/Pk
₹74,200.00INR / Pkg of 25