CPC® (Colder) MPX® Quick-Disconnect Fittings, Cap
Connect to a male coupling insert to stop the flow
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Operate ergonomic thumb latch even with gloved hands
CPC® (Colder) MPX® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Use a cap with an insert to seal off the fluid pathway. Order connectors with a locking sleeve to keep two halves together to guard against accidental disconnection.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
34,100.00 - ₹119,000.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
CPC (Colder) MPX® Quick-Disconnect Fitting, Cap, Polycarbonate; 25/Pk 5011327 Mfr # MPX32003B25
COMPARE
Purple tint Polycarbonate Quick-Disconnect Fitting, Cap, Polycarbonate; 25/Pk
₹34,100.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Cap, Polysulfone; 25/Pk 5011329 Mfr # MPX32039B25
COMPARE
Amber tint Polysulfone Quick-Disconnect Fitting, Cap, Polysulfone; 25/Pk
₹85,700.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Cap with Lock, Polycarbonate; 25/Pk 5011338 Mfr # MPXK32003B25
COMPARE
Purple tint Polycarbonate Quick-Disconnect Fitting, Cap with Lock, Polycarbonate; 25/Pk
₹51,700.00INR / Pkg of 25
CPC (Colder) MPX® Quick-Disconnect Fitting, Cap with Lock, Polysulfone; 25/Pk 5011340 Mfr # MPXK32039B25
COMPARE
Amber tint Polysulfone Quick-Disconnect Fitting, Cap with Lock, Polysulfone; 25/Pk
₹119,000.00INR / Pkg of 25