CPC® (Colder) MPU® Twist-to-Connect Quick-Disconnect Fittings, Hose Barb Insert, Non-Valved, Polysulfone
Join male and female connector halves to quickly create a fluid pathway
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a female coupling body for easy media transfer
  • Easy-to-use twist-to-connect design with locking mechanism
  • 3/4" and 1" hose barbs for high-flow fluid transfer
  • 1" nominal flow path size
The CPC® (Colder) MPU® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. The 3/4" and 1" hose barbs offer fast fluid transfer. The special twist-to-connect design features a locking mechanism that ensures against accidental disconnects. Simply use a cap to seal off the fluid pathway.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
3,800.00 - ₹9,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) MPU® Quick-Disconnect Fitting, Twist-to-Connect, Hose Barb Insert, Polysulfone, Non-Valved, 3/4" ID; 1/Each 5011303 Mfr # MPU221239M
COMPARE
3/4 Polysulfone Quick Disconnect Insert x Hose Barb
₹3,800.00INR / Each
CPC (Colder) MPU® Quick-Disconnect Fitting, Twist-to-Connect, Hose Barb Insert, Polysulfone, Non-Valved, 1" ID; 1/Each 5011304 Mfr # MPU221639M
COMPARE
1 Polysulfone Quick Disconnect Insert x Hose Barb
₹9,700.00INR / Each