CPC® (Colder) MPC™ Quick-Disconnect Fittings, Hose Barb Insert, Non-Valved, Polycarbonate
Connect to a coupling body to quickly create a sterile fluid pathway
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a female coupling body for easy media transfer
  • Mix and match termination sizes for added flexibility
  • 0.375" nominal flow path size
The CPC (Colder) MPC series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Simply use a cap to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
8,800.00 - ₹19,000.00INR / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Buna-N O-Ring, Non-Valved, 1/8" ID; 25/Pk 5011254 Mfr # MPC22002T03B25
COMPARE
1/8 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Buna-N O-Ring, Non-Valved, 1/8" ID; 25/Pk
₹11,800.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Silicone O-Ring, Non-Valved, 1/8" ID; 25/Pk 5011256 Mfr # MPC22002T03MB25
COMPARE
1/8 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Silicone O-Ring, Non-Valved, 1/8" ID; 25/Pk
₹19,000.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Buna-N O-Ring, Non-Valved, 1/4" ID; 25/Pk 5011260 Mfr # MPC22004T03B25
COMPARE
1/4 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Buna-N O-Ring, Non-Valved, 1/4" ID; 25/Pk
₹8,800.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Silicone O-Ring, Non-Valved, 1/4" ID; 25/Pk 5011262 Mfr # MPC22004T03MB25
COMPARE
1/4 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Silicone O-Ring, Non-Valved, 1/4" ID; 25/Pk
₹14,100.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Buna-N O-Ring, Non-Valved, 3/8" ID; 25/Pk 5011266 Mfr # MPC22006T03B25
COMPARE
3/8 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Buna-N O-Ring, Non-Valved, 3/8" ID; 25/Pk
₹8,900.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Silicone O-Ring, Non-Valved, 3/8" ID; 25/Pk 5011268 Mfr # MPC22006T03MB25
COMPARE
3/8 Polycarbonate Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate with Silicone O-Ring, Non-Valved, 3/8" ID; 25/Pk
₹14,000.00INR / Pkg of 25