CPC® (Colder) MPC Quick-Disconnect Fittings, Hose Barb Body, Non-Valved, Polycarbonate
Connect to a coupling insert to quickly create a sterile fluid pathway
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a male coupling inserts for easy media transfer
  • Operate ergonomic thumb latch even with gloved hands
  • Mix and match termination sizes for added flexibility
  • 0.375" nominal flow path size
The CPC (Colder) MPC series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Simply use a plug to seal off the fluid pathway. Order connectors with a locking sleeve to keep two halves together to guard against accidental disconnection. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
14,000.00 - ₹37,900.00INR / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Connection Type Description
Availability Pricing
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Body, Polycarbonate, Non-Valved, 1/8" ID; 25/Pk 5011238 Mfr # MPC17002T03B25
COMPARE
1/8 Quick Disconnect Body x Hose Barb Quick-Disconnect Fitting, Hose Barb Body, Polycarbonate, Non-Valved, 1/8" ID; 25/Pk
₹19,200.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Body, Polycarbonate, Non-Valved, 1/4" ID; 25/Pk 5011242 Mfr # MPC17004T03B25
COMPARE
1/4 Quick Disconnect Body x Hose Barb Quick-Disconnect Fitting, Hose Barb Body, Polycarbonate, Non-Valved, 1/4" ID; 25/Pk
₹14,000.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Body, Polycarbonate, Non-Valved, 3/8" ID; 25/Pk 5011246 Mfr # MPC17006T03B25
COMPARE
3/8 Quick Disconnect Body x Hose Barb Quick-Disconnect Fitting, Hose Barb Body, Polycarbonate, Non-Valved, 3/8" ID; 25/Pk
₹14,000.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Body with Lock, Polycarbonate, Non-Valved, 1/8" ID; 25/Pk 5011286 Mfr # MPCK17002T03B25
COMPARE
1/8 Quick Disconnect Body with Lock x Hose Barb Quick-Disconnect Fitting, Hose Barb Body with Lock, Polycarbonate, Non-Valved, 1/8" ID; 25/Pk
₹37,900.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Body with Lock, Polycarbonate, Non-Valved, 1/4" ID; 25/Pk 5011290 Mfr # MPCK17004T03B25
COMPARE
1/4 Quick Disconnect Body with Lock x Hose Barb Quick-Disconnect Fitting, Hose Barb Body with Lock, Polycarbonate, Non-Valved, 1/4" ID; 25/Pk
₹33,700.00INR / Pkg of 25
CPC (Colder) MPC™ Quick-Disconnect Fitting, Hose Barb Body with Lock, Polycarbonate, Non-Valved, 3/8" ID; 25/Pk 5011294 Mfr # MPCK17006T03B25
COMPARE
3/8 Quick Disconnect Body with Lock x Hose Barb Quick-Disconnect Fitting, Hose Barb Body with Lock, Polycarbonate, Non-Valved, 3/8" ID; 25/Pk
₹33,900.00INR / Pkg of 25