CPC® (Colder) MPC™/MPX® Quick-Disconnect Fittings, Sanitary Body, Non-Valved, Polysulfone
Connect to a coupling insert to quickly create a sterile fluid pathway
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a male coupling insert for easy media transfer
  • Operate ergonomic thumb latch even with gloved hands
  • Mix and match termination sizes for added flexibility
  • 0.375" nominal flow path size
CPC® (Colder) MPC™/MPX® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Simply use a plug to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
3,200.00 - ₹4,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
CPC (Colder) MPC™ Quick-Disconnect Fitting, Sanitary Body, Polysulfone, Non-Valved, 3/4" Sanitary; 1/Each 5011281 Mfr # MPC3301239
COMPARE
Polysulfone Quick Disconnect MPC Body x Sanitary 0.75
₹3,500.00INR / Each
CPC (Colder) MPC™ Quick-Disconnect Fitting, Sanitary Body, Polysulfone, Non-Valved, 1" Sanitary; 1/Each 5011282 Mfr # MPC3301639
COMPARE
Polysulfone Quick Disconnect MPC Body x Sanitary 1
₹4,000.00INR / Each
CPC (Colder) MPX® Quick-Disconnect Fitting, Sanitary Body, Polysulfone, Non-Valved, 3/4" Sanitary; 1/Each 5011330 Mfr # MPX3301239
COMPARE
Polysulfone Quick Disconnect MPX Body x Sanitary 0.75
₹3,200.00INR / Each