CPC® (Colder) MCD Quick-Disconnect Fittings, Hose Barb Body & Insert, Valved, Chrome-Plated Brass
Provides quick and secure connection and disconnection of fluid lines
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly, even with gloved hands
  • Withstands higher pressure and temperature
  • Mix and match termination sizes for added flexibility
  • Compatible with PMC acetal and PMC12 polypropylene couplings
The CPC (Colder) MCD series quick-disconnect hose barb body and insert fittings feature an automatic shutoff valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. Simply push the thumb latch to disconnect. The thumb latch is designed so only one hand is required to safely connect and disconnect the fittings quickly, even with gloves on.
These chrome-plated brass fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures.
3,100.00 - ₹27,200.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) MCD Quick-Disconnect Fitting, Hose Barb Insert, Chrome-Plated Brass, Valved, 1/8" ID; 10/ Pk 3628086 Mfr # MCD2202/10 PK
COMPARE
1/8 Straight Quick-Disconnect Hose Barb Insert, Valve
₹27,200.00INR / Pkg of 10
CPC (Colder) MCD Quick-Disconnect Fitting, Hose Barb Body, Chrome-Plated Brass, Valved, 1/8" ID 5013582 Mfr # MCD1702
COMPARE
1/8 Straight Quick-Disconnect Hose Barb Body, Valve
₹4,700.00INR / Each
CPC (Colder) MCD Quick-Disconnect Fitting, Hose Barb Body, Chrome-Plated Brass, Valved, 1/4" ID 5013583 Mfr # MCD1704
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Valve
₹4,900.00INR / Each
CPC (Colder) MCD Quick-Disconnect Fitting, Hose Barb Insert, Chrome-Plated Brass, Valved, 1/8" ID 5013584 Mfr # MCD2202
COMPARE
1/8 Straight Quick-Disconnect Hose Barb Insert, Valve
₹3,100.00INR / Each
CPC (Colder) MCD Quick-Disconnect Fitting, Hose Barb Insert, Chrome-Plated Brass, Valved, 1/4" ID 5013585 Mfr # MCD2204
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Valve
₹3,200.00INR / Each