CPC® (Colder) LCD Quick-Disconnect Fittings, Hose Barb Body & Insert, Valved, Chrome-Plated Brass
Withstands higher pressures and temperatures
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from ¼” to 3/8” ID
  • Compatible with PLCD acetal and PLCD12 polypropylene couplings
The CPC (Colder) LCD series quick-disconnect hose barb body and insert fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. The high-efficiency design delivers superior flow capacity with minimal spillage in a compact size. An ergonomic thumb latch allows you to safely connect and disconnect the fittings with the use of one hand, even with gloves on.
The fittings can operate in temperatures from -40 to 180 °F (-40 to 82 °C) and pressures from vacuum to 250 psi (17.3 bar).
3,500.00 - ₹5,800.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) LCD Quick-Disconnect Fitting, Hose Barb Body, Chrome-Plated Brass, Valved, 1/4" ID; 1/Each 5013565 Mfr # LCD17004
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Valved
₹5,800.00INR / Each
CPC (Colder) LCD Quick-Disconnect Fitting, Hose Barb Body, Chrome-Plated Brass, Valved, 3/8" ID; 1/Each 5013566 Mfr # LCD17006
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Valved
₹5,600.00INR / Each
CPC (Colder) LCD Quick-Disconnect Fitting, Hose Barb Insert, Chrome-Plated Brass, Valved, 1/4" ID; 1/Each 5013567 Mfr # LCD22004
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Valved
₹3,500.00INR / Each
CPC (Colder) LCD Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Chrome-Plated Brass, Valved, 1/4" ID; 1/Each 5013568 Mfr # LCD23004
COMPARE
1/4 Elbow Quick-Disconnect Hose Barb Insert, Valved
₹3,800.00INR / Each