CPC® (Colder) LC Quick-Disconnect Fittings, Threaded Body & Insert, Non-Valved, Chrome-Plated Brass
Withstands higher pressures and temperatures
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from ¼” to 3/8” ID
  • Compatible with PLC acetal and PLC12 polypropylene couplings
The CPC (Colder) LC series quick-disconnect threaded body and insert fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures. An ergonomic thumb latch allows you to safely connect and disconnect the fittings with the use of one hand, even with gloves on. Easily connect a male insert and a female body to create a quick and secure fluid flow.
The fittings can operate in temperatures from -40 to 180 °F (-40 to 82 °C) and pressures from vacuum to 250 psi (17.3 bar).
1,600.00 - ₹3,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) LC Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Non-Valved, 1/4" NPT(M); 1/Each 5013555 Mfr # LC10004
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valved 1/4
₹3,200.00INR / Each
CPC (Colder) LC Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Non-Valved, 1/4" NPT(M); 1/Each 5013562 Mfr # LC24004
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valved 1/4
₹1,600.00INR / Each
CPC (Colder) LC Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Non-Valved, 3/8" NPT(M); 1/Each 5013563 Mfr # LC24006
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valved 3/8
₹1,800.00INR / Each