CPC® (Colder) LC Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved, Chrome-Plated Brass
Withstands higher pressures and temperatures
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from ¼” to 3/8” ID
  • Compatible with PLC acetal and PLC12 polypropylene couplings
The CPC (Colder) LC series quick-disconnect hose barb body and insert fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures. An ergonomic thumb latch allows you to safely connect and disconnect the fittings with the use of one hand, even with gloves on. Easily connect a male insert and a female body to create a quick and secure fluid flow.
The fittings can operate in temperatures from -40 to 180 °F (-40 to 82 °C) and pressures from vacuum to 250 psi (17.3 bar).
1,500.00 - ₹5,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) LC Quick-Disconnect Fittings, Hose Barb Body, Chrome-Plated Brass, Non-Valved, 1/4" ID; 1/Each 5013556 Mfr # LC17004
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Non-Valved
₹5,800.00INR / Each
CPC (Colder) LC Quick-Disconnect Fittings, Hose Barb Body, Chrome-Plated Brass, Non-Valved, 3/8" ID; 1/Each 5013557 Mfr # LC17006
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Non-Valved
₹5,900.00INR / Each
CPC (Colder) LC Quick-Disconnect Fittings, Hose Barb Insert, Chrome-Plated Brass, Non-Valved, 1/4" ID; 1/Each 5013558 Mfr # LC22004
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Non-Valved
₹1,600.00INR / Each
CPC (Colder) LC Quick-Disconnect Fittings, Hose Barb Insert, Chrome-Plated Brass, Non-Valved, 3/8" ID; 1/Each 5013559 Mfr # LC22006
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Insert, Non-Valved
₹1,500.00INR / Each
CPC (Colder) LC Quick-Disconnect Fittings, Hose Barb Insert, Chrome-Plated Brass, Elbow, Non-Valved, 1/4" ID; 1/Each 5013560 Mfr # LC23004
COMPARE
1/4 Elbow Quick-Disconnect Hose Barb Insert, Non-Valved
₹3,300.00INR / Each
CPC (Colder) LC Quick-Disconnect Fittings, Hose Barb Insert, Chrome-Plated Brass, Elbow, Non-Valved, 3/8" ID; 1/Each 5013561 Mfr # LC23006
COMPARE
3/8 Elbow Quick-Disconnect Hose Barb Insert, Non-Valved
₹3,400.00INR / Each