CPC® (Colder) HFCD12 Quick-Discount Fittings, Threaded Body & Insert, Valved, Polypropylene
High efficiency design provides more flow and lower spillage
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 3/8“ to 3/4” ID
  • Compatible with HFCD35 couplings
The CPC (Colder) HFCD12 Series quick-disconnect threaded insert fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. The high-efficiency design delivers superior flow capacity with minimal spillage in a compact size. Featuring an ergonomic plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation.
Made from chemically resistant polypropylene, they are ideal for harsh environments, providing durability and reliable performance. The fittings can operate in temperatures from 32 to 160 °F (0 to 71 °C) and pressures from vacuum to 60 psi (4.2 bar).
2,300.00 - ₹2,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Threaded Body, Polypropylene, Valved, 1/2" NPT(M); 1/Each 5013533 Mfr # HFCD10812
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 1/2
₹2,900.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, 3/4" NPT(M); 1/Each 5013550 Mfr # HFCD241212
COMPARE
Straight Quick-Disconnect Threaded Insert, Valve 3/4
₹2,300.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, 3/8" NPT(M); 1/Each 5013551 Mfr # HFCD24612
COMPARE
Straight Quick-Disconnect Threaded Insert, Valve 3/8
₹2,300.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, 1/2" NPT(M); 1/Each 5013553 Mfr # HFCD24812
COMPARE
Straight Quick-Disconnect Threaded Insert, Valve 1/2
₹2,300.00INR / Each