CPC® (Colder) HFC12 Quick-Discount Fittings, Threaded Insert, Non-Valved Polypropylene
High efficiency design provides more flow and lower spillage
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 3/8“ to 3/4” ID
  • Compatible with HFC35 couplings
The CPC (Colder) HFC12 Series quick-disconnect threaded insert fittings offer a high-efficiency design, delivering superior flow capacity with minimal spillage in a compact size. Featuring an ergonomic plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation.
Made from chemically resistant polypropylene, they are ideal for harsh environments, providing durability and reliable performance. The fittings can operate in temperatures from 32 to 160 °F (0 to 71 °C) and pressures from vacuum to 60 psi (4.2 bar).
1,100.00 - ₹1,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) HFC12 Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Non-Valved, 3/8" NPT(M); 1/Each 5013530 Mfr # HFC24612
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valve 3/8
₹1,100.00INR / Each
CPC (Colder) HFC12 Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Non-Valved, 1/2" NPT(M); 1/Each 5013532 Mfr # HFC24812
COMPARE
Straight Quick-Disconnect Threaded Insert, Non-Valve 1/2
₹1,200.00INR / Each