CPC® Colder AseptiQuik® W High-Flow Large-Bore Genderless Quick-Disconnect Hose Barb Adapters, Polycarbonate
High-flow, large-bore genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Robust construction provides reliable and repeatable fluid transfer
  • 1.5" nominal flow path size
The AseptiQuik® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, simply unsnap and flip down the protective tab cover, align connectors with the pull tabs hanging down, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
47,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in)
Availability Pricing
CPC (Colder) AseptiQuik® W Quick-Disconnect Fitting, High-Flow Large Bore Genderless Hose Barb, Polycarbonate, 1" ID; 1/Ea 5014363 Mfr # AQW17016
COMPARE
1
₹47,100.00INR / Each
CPC (Colder) AseptiQuik® W Quick-Disconnect Fitting, High-Flow Large Bore Genderless Hose Barb, Polycarbonate, 1-1/4" ID; 1/Ea 5014364 Mfr # AQW17020
COMPARE
1 1/4
₹47,100.00INR / Each
CPC (Colder) AseptiQuik® W Quick-Disconnect Fitting, High-Flow Large Bore Genderless Hose Barb, Polycarbonate, 1-1/2" ID; 1/Ea 5014365 Mfr # AQW17024
COMPARE
1 1/2
₹47,100.00INR / Each