Corning® Round EVA-Foam Ice Buckets with Lid
Recyclable, inert, and nontoxic
  • Highly insulative, unbreakable, lightweight and leakproof
  • Textured finish on bottom prevents slipping on your benchtop
  • Stackable design saves space
  • Keep organized with a variety of bright colors
Made of ethylene-vinyl acetate (EVA), these highly insulative containers will not sweat or leak. They are ideal for use with ice, dry ice (-78°C), liquid nitrogen (-196°C), alcohol or saline solutions, or warm solutions up to 93°C. Lids are included.
23,600.00 - ₹26,400.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Color
Availability Pricing
Corning Ice Bucket, Round 4L, Green, each WW-04393-22 Mfr # 432122
COMPARE
4 Green
₹26,400.00INR / Each
Corning Ice Bucket, Round 4L, Blue, each WW-04393-23 Mfr # 432123
COMPARE
4 Blue
₹26,400.00INR / Each
Corning Ice Bucket, Round 4L, Red, each WW-04393-24 Mfr # 432124
COMPARE
4 Red
₹26,400.00INR / Each
Corning Ice Bucket, Round 4L, Orange, each WW-04393-25 Mfr # 432125
COMPARE
4 Orange
₹26,400.00INR / Each
Corning Ice Bucket, Round 4L, Lime Green, each WW-04393-26 Mfr # 432126
COMPARE
4 Lime Green
₹26,400.00INR / Each
Corning Ice Bucket, Round 4L, Pink, each WW-04393-27 Mfr # 432127
COMPARE
4 Pink
₹26,400.00INR / Each
Corning Ice Bucket, Round 4L, Purple, each WW-04393-28 Mfr # 432128
COMPARE
4 Purple
₹26,400.00INR / Each
Corning Ice Bucket, Round 2.5L, Blue, each WW-04393-29 Mfr # 432129
COMPARE
2.5 Blue
₹23,600.00INR / Each
Corning Ice Bucket, Round 2.5L, Green, each WW-04393-30 Mfr # 432130
COMPARE
2.5 Green
₹23,600.00INR / Each
Corning Ice Bucket, Round 2.5L, Red, each WW-04393-31 Mfr # 432131
COMPARE
2.5 Red
₹23,600.00INR / Each
Corning Ice Bucket, Round 2.5L, Orange, each WW-04393-32 Mfr # 432132
COMPARE
2.5 Orange
₹23,600.00INR / Each
Corning Ice Bucket, Round 2.5L, Lime Green, each WW-04393-33 Mfr # 432133
COMPARE
2.5 Lime Green
₹23,600.00INR / Each
Corning Ice Bucket, Round 2.5L, Pink, each WW-04393-34 Mfr # 432134
COMPARE
2.5 Pink
₹23,600.00INR / Each
Corning Ice Bucket, Round 2.5L, Purple, each WW-04393-35 Mfr # 432135
COMPARE
2.5 Purple
₹23,600.00INR / Each
Showing 1- 14 of 14