Corning® Polypropylene (PP) Centrifuge Tubes
Versatile polypropylene centrifuge tubes for reliable performance across everyday lab workflows
  • Durable polypropylene construction supports a wide range of centrifugation and sample handling applications
  • Multiple cap options including plug seal, CentriStar™, and no-cap configurations to match workflow needs
  • Clear graduations and large marking areas enable accurate measurement and easy sample identification
  • Available in bulk, rack, and universal rack formats across multiple volume sizes
These polypropylene centrifuge tubes are designed to support a broad range of laboratory applications, from routine sample preparation to higher-volume processing. Available in multiple sizes—including 15 mL, 50 mL, 250 mL, and 500 mL—this portfolio offers flexibility to meet varying workflow demands. Durable construction ensures reliable performance during centrifugation, while a range of closure options, including traditional plug seal caps, ergonomic CentriStar™ caps, and no-cap configurations, allows labs to select the right fit for their processes. With multiple packaging formats, including bulk, standard racks, and universal racks, these tubes integrate easily into both high-throughput and organized lab environments.
5,900.00 - ₹117,000.00INR / Pkg of 50
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Style Max RCF x g Packaging
Availability Pricing
Corning 15 mL PP Centrifuge Tubes, Bulk Packed with CentriStar™ Cap, Sterile, 50/Sleeve, 500/Case 4846009 Mfr # 430791
COMPARE
CentriStar 12500 50/Sleeve, 500/Case
₹79,700.00INR / Case of 500
Corning 15 mL PP Centrifuge Tubes, Bulk Packed with No Cap, Sterile, 25/Sleeve, 500/Case 4846010 Mfr # 431470
COMPARE
No cap 12000 25/Sleeve, 500/Case
₹49,600.00INR / Case of 500
Corning 15 mL PP Centrifuge Tubes, Bulk Packed with Plug Seal Cap, Sterile, 50/Sleeve, 500/Case 4846011 Mfr # 430766
COMPARE
Plug seal 12000 50/Sleeve, 500/Case
₹82,700.00INR / Case of 500
Corning 15 mL PP Centrifuge Tubes, Rack Packed with CentriStar™ Cap, Sterile, 50/Rack, 500/Case 4846012 Mfr # 430790
COMPARE
CentriStar 12500 50/Rack, 500/Case
₹82,600.00INR / Case of 500
Corning 15 mL PP Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 50/Rack, 500/Case 4846013 Mfr # 430052
COMPARE
Plug seal 12000 50/Rack, 500/Case
₹86,600.00INR / Case of 500
Corning 250 mL PP Centrifuge Tubes, Plug Seal Cap, Sterile, 6/Pack, 102/Case 4846014 Mfr # 430776
COMPARE
Plug seal 6000 6/Pack, 102/Case
₹77,400.00INR / Case of 102
Corning 50 mL PP Centrifuge Tubes, Bulk Packed with No Cap, Sterile, 25/Sleeve, 500/Case 4846016 Mfr # 431472
COMPARE
No cap 15500 25/Sleeve, 500/Case
₹63,700.00INR / Case of 500
Corning 50 mL PP Centrifuge Tubes, Bulk Packed with CentriStar™ Cap, Sterile, 25/Pack, 500/Case 4846017 Mfr # 430829
COMPARE
CentriStar 17000 25/Pack, 500/Case
₹109,000.00INR / Case of 500
Corning 50 mL PP Centrifuge Tubes, Rack Packed with CentriStar™ Cap, Sterile, 25/Rack, 500/Case 4846018 Mfr # 430828
COMPARE
CentriStar 17000 25/Rack, 500/Case
₹115,000.00INR / Case of 500
Corning 50 mL PP Centrifuge Tubes, Universal Rack Packed with CentriStar™ Cap, Sterile, 25/Universal Rack, 300/Case 4846019 Mfr # 4558
COMPARE
CentriStar 17000 25/Universal Rack, 300/Case
₹71,200.00INR / Case of 300
Corning 50 mL PP Centrifuge Tubes, Bulk Packed with Plug Seal Cap, Sterile, 25/Pack, 500/Case 4846020 Mfr # 430291
COMPARE
Plug seal 15500 25/Pack, 500/Case
₹113,000.00INR / Case of 500
Corning 50 mL PP Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 25/Rack, 500/Case 4846021 Mfr # 430290
COMPARE
Plug seal 15500 25/Rack, 500/Case
₹117,000.00INR / Case of 500
Corning 500 mL PP Centrifuge Tubes, Plug Seal Cap, Sterile, 6/Pack, 36/Case 4846024 Mfr # 431123
COMPARE
Plug seal 6000 6/Pack, 36/Case
₹56,200.00INR / Case of 36
Corning 15 mL PP Centrifuge Tubes, Bulk Packed with CentriStar™ Cap, Sterile, 50/Sleeve 4846040 Mfr # 430791
COMPARE
CentriStar 12500 50/Sleeve
₹9,200.00INR / Pkg of 50
Corning 15 mL PP Centrifuge Tubes, Bulk Packed with No Cap, Sterile, 25/Sleeve 4846041 Mfr # 431470
COMPARE
No cap 12000 25/Sleeve
₹5,900.00INR / Pkg of 25
Corning 15 mL PP Centrifuge Tubes, Bulk Packed with Plug Seal Cap, Sterile, 50/Sleeve 4846042 Mfr # 430766
COMPARE
Plug seal 12000 50/Sleeve
₹9,500.00INR / Pkg of 50
Corning 15 mL PP Centrifuge Tubes, Rack Packed with CentriStar™ Cap, Sterile, 50/Rack 4846043 Mfr # 430790
COMPARE
CentriStar 12500 50/Rack
₹9,500.00INR / Pkg of 50
Corning 15 mL PP Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 50/Rack 4846044 Mfr # 430052
COMPARE
Plug seal 12000 50/Rack
₹10,000.00INR / Pkg of 50
Corning 250 mL PP Centrifuge Tubes, Plug Seal Cap, Sterile, 6/Pack 4846045 Mfr # 430776
COMPARE
Plug seal 6000 6/Pack
₹8,800.00INR / Pkg of 6
Corning 50 mL PP Centrifuge Tubes, Bulk Packed with No Cap, Sterile, 25/Sleeve 4846047 Mfr # 431472
COMPARE
No cap 15500 25/Sleeve
₹7,400.00INR / Pkg of 25
Showing 1 - 20 of 26