Corning® Polypropylene (PP) Centrifuge Tubes, Self-Standing
Self-standing 50 mL centrifuge tubes for stable handling
  • Self-standing base improves stability during sample prep, transport, and short-term storage
  • Supports reliable centrifugation up to ~3,000 × g for standard lab applications
  • Traditional plug seal for a secure closure or CentriStar™ caps for easier grip and handling
  • Printed graduations and large marking areas simplify measurement and sample identification
These self-standing 50 mL centrifuge tubes are designed for convenience and control during everyday lab work. Their flat-bottom design allows tubes to remain upright without external support, reducing the need for racks and improving workflow efficiency during sample preparation and handling. Made from durable polypropylene and available with either standard plug seal or ergonomic CentriStar™ caps, these tubes provide a practical, easy-to-use solution for labs prioritizing stability, organization, and reliable routine performance.
13,200.00 - ₹123,000.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Cap Style Packaging
Availability Pricing
Corning 50 mL PP Centrifuge Tubes, Self-Standing with CentriStar™ Cap, Bulk Packed, Sterile, 25/Pack, 500/Case 4846022 Mfr # 430921
COMPARE
CentriStar 25/Pack, 500/Case
₹119,000.00INR / Case of 500
Corning 50 mL PP Centrifuge Tubes, Self-Standing with Plug Seal Cap, Bulk Packed, Sterile, 25/Pack, 500/Case 4846023 Mfr # 430897
COMPARE
Plug seal 25/Pack, 500/Case
₹123,000.00INR / Case of 500
Corning 50 mL PP Centrifuge Tubes, Self-Standing with CentriStar™ Cap, Bulk Packed, Sterile, 25/Pack 4846053 Mfr # 430921
COMPARE
CentriStar 25/Pack
₹13,200.00INR / Pkg of 25
Corning 50 mL PP Centrifuge Tubes, Self-Standing with Plug Seal Cap, Bulk Packed, Sterile, 25/Pack 4846054 Mfr # 430897
COMPARE
Plug seal 25/Pack
₹13,700.00INR / Pkg of 25