Corning® Polyethylene Terephthalate (PET) Centrifuge Tubes
High-clarity PET centrifuge tubes for precise sample handling and dependable results
  • PET construction supports centrifugation up to 3,600 × g
  • DNase-/RNase-free and nonpyrogenic to protect sensitive samples
  • Clear tubes with printed graduations and large marking areas for accurate measurement and labeling
  • 95 k PA (14 psi) pressure tested
These 15 mL PET centrifuge tubes provide dependable performance for everyday lab workflows while giving you flexibility in how you store and handle samples. Both options feature sterile, plug-seal designs with clear graduations. Choose bulk packaging to maximize storage efficiency and reduce packaging waste, or opt for racked tubes to streamline workflow and improve bench organization.
9,500.00 - ₹137,000.00INR / Case of 500
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Packaging Tube Capacity (mL)
Availability Pricing
Corning 15 mL PET Centrifuge Tubes, Bulk Packed with Plug Seal Cap, Sterile, 50/Sleeve, 500/Case 4846007 Mfr # 430053
COMPARE
50/Sleeve, 500/Case 15
₹82,700.00INR / Case of 500
Corning 15 mL PET Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 50/Rack, 500/Case 4846008 Mfr # 430055
COMPARE
50/Rack, 500/Case 15
₹87,300.00INR / Case of 500
Corning 50 mL PET Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 25/Rack, 500/Case 4846015 Mfr # 430304
COMPARE
25/Rack, 500/Case 50
₹137,000.00INR / Case of 500
Corning 15 mL PET Centrifuge Tubes, Bulk Packed with Plug Seal Cap, Sterile, 50/Sleeve 4846038 Mfr # 430053
COMPARE
50/Sleeve 15
₹9,500.00INR / Pkg of 50
Corning 15 mL PET Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 50/Rack 4846039 Mfr # 430055
COMPARE
50/Rack 15
₹10,000.00INR / Pkg of 50
Corning 50 mL PET Centrifuge Tubes, Rack Packed with Plug Seal Cap, Sterile, 25/Rack 4846046 Mfr # 430304
COMPARE
25/Rack 50
₹15,300.00INR / Pkg of 25