Corning® CoolSink® Plate and Reservoir Modules
Rapid temperature adaption to any temperature source
  • Reduce temperature variability and uncertainty with uniform well-to-well temperature
  • Ensured temperature accuracy of ±0.1° C across all tubes and plates for critical applications
  • Easy to clean or sterilize
These thermo-conductive plate and reservoir modules provide uniform temperature to all wells when cooling, snap freezing, heating, or thawing samples. Ideal for use in a variety of temperature sources such as ice, dry ice, liquid nitrogen, or water baths. The modules will rapidly adapt to the temperature of the source being used, from -196°C to >100°C. All options may be autoclaved, high heat sterilized, or decontaminated with bleach, alcohol, or other disinfectants or lab detergents. Both plate modules are SBS-compatible.
Use in traditional ice buckets and pans, or for an ice-free option, use with Corning CoolBox ice-free cooling systems.
124,000.00 - ₹139,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity Description
Availability Pricing
Corning XT CoolSink 96F , each 0439270 Mfr # 432070
COMPARE
One 6-, 12-, 24-, 48-, 96-, or 384-well flat-bottom plate CoolSink XT 96F
₹124,000.00INR / Each
Corning XT CoolSink 96U, each 0439271 Mfr # 432071
COMPARE
One 96-well round-bottom microplate CoolSink XT 96U
₹139,000.00INR / Each