Cooper-Atkins DFP450W Waterproof digital thermometer with reduced tip and temperature alarm.
Ideal for use in the food industry and all other general applications
  • NSF approved
Antimicrobial protection added to all plastic components. Thermometer is factory calibrated to three temperature points and guaranteed to never need field adjustment. Features include IP67 rating, stainless steel shaft, 6-second response time, °F/°C switchable ranges, hold function, min/max reading, and auto-off. Plus the addition of a unique adjustable alarm function. Dishwasher-safe.
14,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
Cooper-Atkins DFP450W-0-8 Waterproof Reduced-Tip Digital Thermometer; -40 to 450 F 9002501 Mfr # DFP450W-0-8
COMPARE
450 Digital Indicator 1
₹14,000.00INR / Each