Contec EasySat Bucketless Floor Mop Heads
EasySat™ Mop heads are made from ultra-clean polyester/ester foam VertiKlean material. Available with flat or textured surface and standard or gamma irradiated to 10-6 Sterility Assurance Level per AAMI/ISO 11137:2006 guidelines.
70,800.00 - ₹73,500.00INR / Pkg of 32
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Contec EasySat Mop Head for Bucketless Floor Mop WW-09411-31 Mfr # BLVKMOP
COMPARE
Mop Head for Bucketless Floor Mop
₹70,800.00INR / Pkg of 32
Contec EasySat Textured Mop Head for Bucketless Floor Mop WW-09411-35 Mfr # TBLVKMOP
COMPARE
Textured Mop Head for Bucketless Floor Mop
₹73,500.00INR / Pkg of 32