Contact/Laser Tachometers
Compact, economical units are packed with features
  • Multiple units of measure for rate and total revolutions – no conversion necessary
  • Large LCD is easy to read with units of measure indicated
These rugged tachometers fit perfectly in the left or right hand, and are designed for use with a gloved hand. Units provide fourteen different units of measure including rpm, inch/minute, feet/minute to total length measurements like feet and meters, and more. Additional memory features include last, minimum, maximum, and average of results. Durable units are ideal for the most challenging industrial environments, and the compact design is small enough to fit into your shirt pocket.
Model 98807-20 noncontact tachometer is easy to use and incorporates a laser that is accurate up to 6 ft (1.8 m) from the target, ensuring a safe distance from rotating parts.
61,000.00 - ₹71,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max RPM Range (rpm)
Availability Pricing
Shimpo MT-100 Compact Contact Pocket/Handheld Tachometer 9880719 Mfr # MT-100
COMPARE
5-digit LCD 25000
₹61,000.00INR / Each
Shimpo MT-200 Digital Laser Compact Pocket Hand Tachometer 9880720 Mfr # MT-200
COMPARE
5-digit LCD 99999
₹71,700.00INR / Each