Cone Bottom Tanks
Tanks are ideal for small mix and batch applications
  • Available in 30 and 45 degree slopes
  • Specific gravity ratings are based on the industry's most severe calculation
Small to medium size batch tanks save space and are perfect for smaller and short-term mixing, batch processes, and liquid storage. Cone bottom tanks are constructed high-density linear polyethylene and have a maximum storage temperature of 140°F. Specific gravity of tanks is 1.9. Material is UV stabilized for long term outdoor service. Material is FDA compliant and meets NSF standard 61. Model 06960-26 comes with a 17”W removable cover. All others come with 10”W caps. Manufactured with stratiform all thickness- walls are thicker at bottom of tank. Stands are constructed of steel and allow a nominal bottom clearance of 12” for total drain capability. Order stands seperately.
71,300.00 - ₹74,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Temperature (° F) Volume (Gal)
Availability Pricing
Snyder 1520000N45 Cone bottom tank, 15 gallons, 45° cone 0696026 Mfr # 1520000N45
COMPARE
HDLPE 140 15
₹74,900.00INR / Each
Snyder 1580000N45 Cone bottom tank, 35 gallons, 30° cone 0696028 Mfr # 1580000N45
COMPARE
HDLPE 140 35
₹71,300.00INR / Each
Snyder 1560000N45 Cone bottom tank, 65 gallons, 30° cone 0696030 Mfr # 1560000N45
COMPARE
HDLPE 140 65
₹74,300.00INR / Each