Compression to NPT(M) Adapter Fittings, PP, 10/pack
For water-treatment, DI water, and mild chemical-feed tubing connections
  • 20% glass-filled polypropylene offers good chemical resistance and improved strength
  • Ideal for water-treatment, filtration, DI water, and mild chemical-feed connections
  • Unaffected by alkalis, salts, and most weak acids
  • Material temperature rating: -34 to 107°C (-30 to 225°F)
  • Plastic gripper nut helps secure plastic tubing
  • NSF 14 certified, NSF 61 compliant, and autoclavable
JACO polypropylene compression to male thread adapters connect rigid plastic tubing to threaded equipment ports in water-treatment, filtration, deionized-water, and mild chemical-feed systems. Molded from 20% glass-filled polypropylene, these fittings are lightweight and rigid, with improved strength, resistance to environmental stress cracking, and good chemical resistance.
Polypropylene is a dependable choice for applications involving alkalis, salts, and most weak acids, including hydrochloric and phosphoric acids. Below 79°C (175°F), it also offers good resistance to organic solvents. Its inert properties make it suitable for high-purity applications such as deionized water, where polypropylene compatibility and required system certifications have been verified.
These fittings have a solid opaque, flat white appearance. Polypropylene has a material temperature rating of -34 to 107°C (-30 to 225°F). JACO polypropylene fittings are NSF 14 certified, NSF 61 compliant, and autoclavable.
2,700.00 - ₹4,400.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/4 1/4
₹2,700.00INR / Pkg of 10
COMPARE
3/8 3/8
₹3,500.00INR / Pkg of 10
COMPARE
1/2 1/2
₹4,400.00INR / Pkg of 10