Compact Angle Pattern 2-Way Pressure Relief/Bypass Valves
Ensure safety with automatic release of pressure
  • Quick response, single-flat diaphragm prevents crystallization
  • PTFE spring guides provide smooth, chatter-free operation
  • Machined from solid thermoplastic for durability
  • Infinitely adjustable pressure relief setting from 5 to 150 psi
These angle pattern pressure relief/bypass valves feature a single diaphragm that delivers a quick response and prevents crystallization buildup that can lead to sticking. Easily set an infinitely adjustable relief setting from 5 to 150 psi; valves handle inlet pressures up to 210 psi. Precision-engineered PTFE guides ensure smooth, chatter-free operation.
Machined from solid thermoplastics, these valves withstand harsh, corrosive fluids and offer excellent durability. Use these versatile valves for pressure relief, back pressure regulation, bypass relief, and pump back pressure control applications to help protect systems, maintain consistent pressure, and extend pump life.
15,800.00 - ₹251,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Housing Material Inlet Port Outlet Port
Availability Pricing
Compact Angle Pattern 2-Way Pressure Relief/Bypass Valve, PP, 1/4" NPT(F) 4852717 Mfr # RVD025V-PP
COMPARE
Polypropylene 1/4" NPT(F) 1/4" NPT(F)
₹15,800.00INR / Each
Compact Angle Pattern 2-Way Pressure Relief/Bypass Valve, PVC, 1/4" NPT(F) 4852718 Mfr # RVD025V-PV
COMPARE
PVC 1/4" NPT(F) 1/4" NPT(F)
₹92,900.00INR / Each
Compact Angle Pattern 2-Way Pressure Relief/Bypass Valve, PTFE, 1/4" NPT(F) 4852719 Mfr # RVD025V-TF
COMPARE
PTFE 1/4" NPT(F) 1/4" NPT(F)
₹229,000.00INR / Each
Compact Angle Pattern 2-Way Pressure Relief/Bypass Valve, PP, 1/2" NPT(F) 4852720 Mfr # RVD050V-PP
COMPARE
Polypropylene 1/2" NPT(F) 1/2" NPT(F)
₹125,000.00INR / Each
Compact Angle Pattern 2-Way Pressure Relief/Bypass Valve, PVC, 1/2" NPT(F) 4852721 Mfr # RVD050V-PV
COMPARE
PVC 1/2" NPT(F) 1/2" NPT(F)
₹103,000.00INR / Each
Compact Angle Pattern 2-Way Pressure Relief/Bypass Valve, PTFE, 1/2" NPT(F) 4852722 Mfr # RVD050V-TF
COMPARE
PTFE 1/2" NPT(F) 1/2" NPT(F)
₹251,000.00INR / Each