Combustible Cabinets
Reduce fire risks and protect workers
  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
Use to store paints, oils, and other flammable liquids (high flash point liquid, flash point ≥37.8). Suitable for coating, printing, furniture, automobile, and other industries.
Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and another on the bottom) effectively reduce the concentration of volatile substances and prevent fire hazards. Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.
46,400.00 - ₹148,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Number of Shelves Description
Availability Pricing
SYSBEL WA810040R 4 Gal/15L,1 door, manual 1597679 Mfr # WA810040R
COMPARE
15 1 Combustible Cabinets; 4 Gal/15L,1 door, manual
₹46,400.00INR / Each
SYSBEL WA810120R 12 Gal/45L,1 door, manual 1597680 Mfr # WA810120R
COMPARE
45 1 Combustible Cabinets; 12 Gal/45L,1 door, manual
₹64,400.00INR / Each
SYSBEL WA810300R 30 Gal/114L,2 door, manual 1597681 Mfr # WA810300R
COMPARE
114 1 Combustible Cabinets; 30 Gal/114L,2 door, manual
₹97,700.00INR / Each
SYSBEL WA810450R 45 Gal/170L,2 door, manual 1597682 Mfr # WA810450R
COMPARE
170 2 Combustible Cabinets; 45 Gal/170L,2 door, manual
₹123,000.00INR / Each
SYSBEL WA810600R 60 Gal/227L,2 door, manual 1597683 Mfr # WA810600R
COMPARE
227 1 Combustible Cabinets; 60 Gal/227L,2 door, manual
₹137,000.00INR / Each
SYSBEL WA810860R 90 Gal/340L,2 door, manual 1597684 Mfr # WA810860R
COMPARE
340 2 Combustible Cabinets; 90 Gal/340L,2 door, manual
₹148,000.00INR / Each